Assay Details
| Gene Symbol: Cpeb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252525.1 | NP_001239454.1 | 10 - 11 | 1621 | 92 | |
| NM_001252526.1 | NP_001239455.1 | 10 - 11 | 1603 | 92 | ||
| NM_007755.5 | NP_031781.1 | 10 - 11 | 1618 | 92 | ||
| GenBank mRNA | AK136088.1 | - | 10 - 11 | 1622 | 92 | |
| BC125476.1 | - | 10 - 11 | 1538 | 40129991 | 92 | |
| BC144948.1 | - | 10 - 11 | 1520 | 92 | ||
| Y08260.1 | - | 10 - 11 | 1537 | 92 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cpeb1
Genomic Map