Assay Details
| Gene Symbol: Ccdc122 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175369.3 | NP_780578.1 | 1 - 2 | 198 | 59 | |
| GenBank mRNA | AK077161.1 | - | 1 - 2 | 199 | 59 | |
| BC096577.1 | - | 1 - 2 | 192 | 59 | ||
| BC138881.1 | - | 1 - 2 | 106 | 8861903 | 59 | |
| BC138883.1 | - | 1 - 2 | 106 | 8861905 | 59 | |
| BC145455.1 | - | 1 - 2 | 106 | 59 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK077161.1, BC096577.1, BC138881.1, BC138883.1, NM_175369.3 transcripts.
Set Membership: |
|

Ccdc122
Genomic Map