Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700010254.mm.1 |
TC0700001168.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700010254.mm.1 |
NM_001004184.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700010254.mm.1 |
NM_001004184.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700010255.mm.1 |
TC0700001168.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700010255.mm.1 |
NM_001004184.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700010255.mm.1 |
NM_001004184.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700010257.mm.1 |
TC0700001168.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700010257.mm.1 |
NM_001004184.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700010257.mm.1 |
NM_001004184.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700010258.mm.1 |
TC0700001168.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700010258.mm.1 |
NM_001004184.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700010258.mm.1 |
NM_001004184.3 |
Refseq |