Assay Details
| Gene Symbol: Whamm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004185.3 | NP_001004185.3 | 2 - 3 | 793 | 74 | |
| GenBank mRNA | AK080911.1 | - | 3 - 4 | 526 | 74 | |
| AK162084.1 | - | 4 - 5 | 798 | 74 | ||
| AK220273.1 | - | 2 - 3 | 793 | 74 | ||
| BC042749.2 | - | 3 - 4 | 628 | 5322109 | 74 | |
| BC050804.1 | - | 2 - 3 | 256 | 6771347 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Whamm
Genomic Map