Assay Details
| Gene Symbol: Dpysl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009955.3 | NP_034085.2 | 1 - 2 | 268 | 83 | |
| GenBank mRNA | BC062955.1 | - | 1 - 2 | 267 | 30535984 | 83 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC062955.1, NM_009955.3 transcripts.
Set Membership: |
|

Dpysl2
Genomic Map