Assay Details
| Gene Symbol: Dpysl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009955.3 | NP_034085.2 | 6 - 7 | 850 | 68 | |
| GenBank mRNA | BC062955.1 | - | 6 - 7 | 849 | 30535984 | 68 |
More Information
Updated Mapping Information
Set Membership: |
|

Dpysl2
Genomic Map