Assay Details
| Gene Symbol: Ang4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AY219870.1 | - | 2 | 437 | 100 | |
| BC042938.1 | - | 1 | 367 | 6398297 | 100 | |
| BC132444.1 | - | 2 | 390 | 40130721 | 100 | |
| BC132448.1 | - | 2 | 390 | 40130725 | 100 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ang4
Genomic Map