Assay Details
| Gene Symbol: 4930570G19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040399.1 | 2 - 3 | 416 | 114 | ||
| GenBank mRNA | AK016265.1 | - | 2 - 3 | 416 | 114 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 9330178D15 gene | 9330178D15Rik | 9330178D15 | -539.0kb | Anti-sense | Chr. 3 - 155976354 - 156008592 on NCBI Build GRCm38 |
| Downstream | neuronal growth regulator 1 | Negr1 |
5330422G01Rik, KILON, Ntra |
10.6kb | Anti-sense | Chr. 3 - 156561794 - 157316464 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

4930570G19Rik
Genomic Map