| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix HT MG-430 PM Array Plate |
1439684_PM_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1439684_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1439684_PM_at |
- |
- |
| Affymetrix Mouse Clariom S Array |
PSR0300024990.mm.1 |
NR_040399.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300024990.mm.1 |
NR_040399.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300024990.mm.1 |
TC0300003187.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300024989.mm.1 |
NR_040399.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300024989.mm.1 |
NR_040399.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300024989.mm.1 |
TC0300003187.mm.1 |
MTA TC |
| Mouse Expression Set 430 |
1439684_PM_at |
- |
- |
| Mouse Expression Set 430 |
1439684_at |
- |
- |
| Murine Genome U74v2 Set |
136175_at |
- |
- |
| Murine Genome U74v2 Set |
136175_PM_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1439684_PM_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1439684_at |
- |
- |