Assay Details
Gene Symbol: 1700012D16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK005899.1 | - | 1 - 2 | 79 | 77 | |
BY706039.1 | - | 1 - 2 | 79 | 77 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | ST6 (alpha-N-acetyl-neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminide alpha-2,6-sialyltransferase 3 | St6galnac3 | Siat7c | Sense | Chr. 3 - 153202509 - 153725133 on NCBI Build GRCm38 | |
Downstream | RIKEN cDNA 5730460C07 gene | 5730460C07Rik | 490.7kb | Sense | Chr. 3 - 153789305 - 153792087 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
