Assay Details
| Gene Symbol: Efcab2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026626.2 | NP_080902.1 | 4 - 5 | 497 | 68 | |
| GenBank mRNA | AK010880.1 | - | 4 - 5 | 350 | 68 | |
| AK015157.1 | - | 4 - 5 | 279 | 68 | ||
| AK018894.1 | - | 4 - 5 | 358 | 68 | ||
| AK019663.1 | - | 4 - 5 | 290 | 68 | ||
| AK019802.1 | - | 4 - 5 | 299 | 68 | ||
| BC025062.1 | - | 4 - 5 | 497 | 4240831 | 68 | |
More Information
Set Membership: |
|

Efcab2
Genomic Map