Assay Details
| Gene Symbol: Nfkb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008689.2 | NP_032715.2 | 11 - 12 | 108 | ||
| GenBank mRNA | AB119195.1 | - | 9 - 10 | 911 | 108 | |
| AB221614.1 | - | 9 - 10 | 108 | |||
| AK052726.1 | - | 6 - 7 | 108 | |||
| AY521462.1 | - | 9 - 10 | 911 | 108 | ||
| AY521463.1 | - | 9 - 10 | 108 | |||
| BC050841.1 | - | 11 - 12 | 108 | |||
| BC138535.1 | - | 10 - 11 | 8861555 | 108 | ||
| BC138536.1 | - | 10 - 11 | 8861556 | 108 | ||
| BC145317.1 | - | 11 - 12 | 108 | |||
| M57999.1 | - | 11 - 12 | 1202 | 108 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nfkb1
Genomic Map