Assay Details
| Gene Symbol: Nfkb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008689.2 | NP_032715.2 | 1 - 2 | 424 | 68 | |
| GenBank mRNA | BC050841.1 | - | 1 - 2 | 424 | 68 | |
| M57999.1 | - | 1 - 2 | 67 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC050841.1, M57999.1, NM_008689.2 transcripts.
Set Membership: |
|

Nfkb1
Genomic Map