Assay Details
| Gene Symbol: Nfkb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008689.2 | NP_032715.2 | 23 - 24 | 3082 | 71 | |
| GenBank mRNA | AB119195.1 | - | 21 - 22 | 2580 | 71 | |
| AB221614.1 | - | 21 - 22 | 2587 | 71 | ||
| AK089660.1 | - | 11 - 12 | 1456 | 71 | ||
| AK146389.1 | - | 9 - 10 | 1301 | 71 | ||
| AK157915.1 | - | 8 - 9 | 1082 | 71 | ||
| AK159423.1 | - | 11 - 12 | 1513 | 71 | ||
| AY521462.1 | - | 21 - 22 | 2580 | 71 | ||
| AY521463.1 | - | 21 - 22 | 2580 | 71 | ||
| BC138535.1 | - | 22 - 23 | 2659 | 8861555 | 71 | |
| BC138536.1 | - | 22 - 23 | 2659 | 8861556 | 71 | |
| BC145317.1 | - | 23 - 24 | 2676 | 71 | ||
| M57999.1 | - | 23 - 24 | 2871 | 71 | ||
| S66656.1 | - | 11 - 12 | 1626 | 71 | ||
| S89033.1 | - | 11 - 12 | 1674 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nfkb1
Genomic Map