Assay Details
| Gene Symbol: Nfkb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008689.2 | NP_032715.2 | 3 - 4 | 535 | 71 | |
| GenBank mRNA | BC050841.1 | - | 3 - 4 | 535 | 71 | |
| BC138535.1 | - | 2 - 3 | 112 | 8861555 | 71 | |
| BC138536.1 | - | 2 - 3 | 112 | 8861556 | 71 | |
| BC145317.1 | - | 2 - 3 | 112 | 71 | ||
| M57999.1 | - | 3 - 4 | 324 | 71 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC050841.1, BC138535.1, BC138536.1, M57999.1, NM_008689.2 transcripts.
Set Membership: |
|

Nfkb1
Genomic Map