Assay Details
| Gene Symbol: Tcp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290712.1 | NP_001277641.1 | 1 - 2 | 301 | 110 | |
| GenBank mRNA | AK151445.1 | - | 1 - 2 | 191 | 110 | |
| AK153430.1 | - | 1 - 2 | 191 | 110 | ||
| CN725473.1 | - | 1 - 2 | 301 | 110 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK151445.1, AK153430.1, NM_001290712.1 transcripts.
Set Membership: |
|

Tcp1
Genomic Map