Assay Details
| Gene Symbol: Ncoa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010881.2 | NP_035011.1 | 14 - 15 | 3439 | 103 | |
| GenBank mRNA | BC068177.1 | - | 11 - 12 | 2747 | 30628581 | 103 |
| BC080866.1 | - | 14 - 15 | 3202 | 30532122 | 103 | |
| U56920.1 | - | 13 - 14 | 2997 | 103 | ||
| U64606.1 | - | 11 - 12 | 2742 | 103 | ||
| U64828.1 | - | 14 - 15 | 3352 | 103 | ||
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | AK089302.1 | - | Chr. - - [-] on Build | 103 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Ncoa1
Genomic Map