Assay Details
| Gene Symbol: Ncoa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010881.2 | NP_035011.1 | 17 - 18 | 4035 | 56 | |
| GenBank mRNA | BC068177.1 | - | 14 - 15 | 3343 | 30628581 | 56 |
| BC080866.1 | - | 17 - 18 | 3798 | 30532122 | 56 | |
| U56920.1 | - | 16 - 17 | 3593 | 56 | ||
| U64606.1 | - | 14 - 15 | 3338 | 56 | ||
| U64828.1 | - | 17 - 18 | 3948 | 56 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ncoa1
Genomic Map