Assay Details
| Gene Symbol: Ncoa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010881.2 | NP_035011.1 | 18 - 19 | 4438 | 85 | |
| GenBank mRNA | AK035922.1 | - | 3 - 4 | 762 | 85 | |
| BC068177.1 | - | 15 - 16 | 3746 | 30628581 | 85 | |
| BC080866.1 | - | 18 - 19 | 4201 | 30532122 | 85 | |
| U56920.1 | - | 17 - 18 | 3996 | 85 | ||
| U64606.1 | - | 15 - 16 | 3741 | 85 | ||
| U64828.1 | - | 18 - 19 | 4351 | 85 | ||
More Information
Set Membership: |
|

Ncoa1
Genomic Map