Assay Details
| Gene Symbol: Mpc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018819.4 | NP_061289.1 | 1 - 3 | 151 | 123 | |
| GenBank mRNA | AF181116.1 | - | 1 - 3 | 107 | 123 | |
| AK002241.1 | - | 1 - 3 | 152 | 123 | ||
| AK002266.1 | - | 1 - 3 | 130 | 123 | ||
| AK004013.1 | - | 1 - 3 | 107 | 123 | ||
| AK005348.1 | - | 1 - 3 | 145 | 121 | ||
| AK013391.1 | - | 1 - 3 | 152 | 123 | ||
| AK014421.1 | - | 1 - 3 | 109 | 123 | ||
| AK135943.1 | - | 1 - 3 | 122 | 123 | ||
| AK170514.1 | - | 1 - 3 | 168 | 123 | ||
| BC024365.1 | - | 1 - 3 | 113 | 4988147 | 123 | |
| BC103767.1 | - | 1 - 3 | 154 | 30840199 | 123 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mpc1
Genomic Map