Assay Details
| Gene Symbol: Kif3c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008445.2 | NP_032471.2 | 6 - 7 | 2974 | 141 | |
| GenBank mRNA | AF013116.1 | - | 6 - 7 | 2550 | 141 | |
| AK139894.1 | - | 6 - 7 | 650 | 141 | ||
| AK147572.1 | - | 6 - 7 | 2776 | 141 | ||
| AK220283.1 | - | 7 - 8 | 2699 | 141 | ||
| BC052732.1 | - | 6 - 7 | 2131 | 141 | ||
| BC053053.1 | - | 6 - 7 | 2916 | 141 | ||
| BC062648.1 | - | 6 - 7 | 2649 | 141 | ||
| BC127063.1 | - | 6 - 7 | 2186 | 40109965 | 141 | |
More Information
Updated Mapping Information
Set Membership: |
|

Kif3c
Genomic Map