Assay Details
Gene Symbol: Dmpk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190490.1 | NP_001177419.1 | 9 - 10 | 1216 | 110 | |
NM_001190491.1 | NP_001177420.1 | 8 - 9 | 1294 | 110 | ||
NM_032418.2 | NP_115794.1 | 8 - 9 | 1294 | 110 | ||
GenBank mRNA | BC024150.1 | - | 8 - 9 | 1281 | 4217643 | 110 |
BC075715.1 | - | 9 - 10 | 1124 | 4989259 | 110 | |
S60313.1 | - | 5 - 6 | 801 | 110 | ||
S60314.1 | - | 5 - 6 | 801 | 110 | ||
S60315.1 | - | 6 - 7 | 714 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
