Assay Details
| Gene Symbol: Styx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019637.3 | NP_062611.2 | 3 - 4 | 431 | 101 | |
| GenBank mRNA | AK002822.1 | - | 373 | 101 | ||
| AK030122.1 | - | 3 - 4 | 431 | 101 | ||
| AK046970.1 | - | 3 - 4 | 408 | 101 | ||
| AK154136.1 | - | 3 - 4 | 526 | 101 | ||
| AK166043.1 | - | 3 - 4 | 414 | 101 | ||
| BC038608.1 | - | 3 - 4 | 381 | 5012303 | 101 | |
| BC085089.1 | - | 3 - 4 | 378 | 6492329 | 101 | |
| U34973.1 | - | 3 - 4 | 399 | 101 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Styx
Genomic Map