Assay Details
| Gene Symbol: Styx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019637.3 | NP_062611.2 | 8 - 9 | 718 | 115 | |
| GenBank mRNA | AK002822.1 | - | 660 | 115 | ||
| AK030122.1 | - | 8 - 9 | 718 | 115 | ||
| AK042741.1 | - | 9 - 10 | 773 | 115 | ||
| AK046970.1 | - | 8 - 9 | 695 | 115 | ||
| AK166043.1 | - | 8 - 9 | 701 | 115 | ||
| BC038608.1 | - | 8 - 9 | 668 | 5012303 | 115 | |
| BC096674.1 | - | 7 - 8 | 628 | 5144003 | 115 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Styx
Genomic Map