Assay Details
| Gene Symbol: Cdkn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028222.1 | NP_082498.1 | 4 - 5 | 289 | 90 | |
| GenBank mRNA | AK010426.1 | - | 4 - 5 | 247 | 90 | |
| AK033341.1 | - | 4 - 5 | 253 | 90 | ||
| AK146312.1 | - | 4 - 5 | 205 | 90 | ||
| BC049694.1 | - | 4 - 5 | 273 | 6705916 | 90 | |
| BY084534.1 | - | 4 - 5 | 288 | 90 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29578140 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29578139 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29578141 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29578142 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29578138 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29578137 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P30989 | AK033341.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P30989 | AK146312.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P30989 | AK010426.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P30989 | BC049694.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P30989 | NM_028222.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P30989 | AK033341.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P30989 | AK146312.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P30989 | AK010426.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P30989 | BC049694.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P30989 | NM_028222.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 216816 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 441102 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 947606 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 602838 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 370537 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 436455 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 419699 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 142589 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P128305 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P136164 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P30989 | AK033341.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P30989 | AK146312.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P30989 | AK010426.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P30989 | BC049694.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P30989 | NM_028222.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P30989 | AK033341.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P30989 | AK146312.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P30989 | AK010426.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P30989 | BC049694.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P30989 | NM_028222.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P128305 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P136164 | - | - |
| Whole Mouse Genome G4122A | A_52_P30984 | BC049694.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P30984 | NM_028222.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P285651 | AK033341.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P30989 | AK033341.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P30989 | AK146312.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P30989 | AK010426.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P30989 | BC049694.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P30989 | NM_028222.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P30989 | AK033341.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P30989 | AK146312.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P30989 | AK010426.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P30989 | BC049694.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P30989 | NM_028222.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P128305 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P136164 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29578137 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29578139 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29578138 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29578140 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29578142 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29578141 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29578175 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11825692 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29578123 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29578111 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29578136 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29578180 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29578170 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29578150 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29578133 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29578177 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29578158 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11586792 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29578154 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11284008 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29578178 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29578147 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29578165 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11611919 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11711521 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29578137 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29578162 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11835195 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29578143 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29578169 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11532275 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29578149 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29578115 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11284001 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29578120 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29578110 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29578124 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29578114 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29578126 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11835193 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29578112 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29578155 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11284017 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29578132 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29578160 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29578151 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29578117 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29578172 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29578167 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29578164 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29578139 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29578118 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29578157 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29578130 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10414318 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10414323 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10414319 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10414324 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10414317 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10414322 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10414316 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10414320 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10414321 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1430574_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10414315 | - | - |
| Mouse Genome 430 2.0 Array | 1430574_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1430574_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 72391_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 72391_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1400004062.mm.1 | NM_028222.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400004062.mm.1 | NM_028222.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400004062.mm.1 | TC1400000501.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1400004064.mm.1 | NM_028222.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400004064.mm.1 | NM_028222.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400004064.mm.1 | TC1400000501.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1400004065.mm.1 | NM_028222.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400004065.mm.1 | NM_028222.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400004065.mm.1 | TC1400000501.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1400004066.mm.1 | NM_028222.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400004066.mm.1 | NM_028222.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400004066.mm.1 | TC1400000501.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1430574_PM_at | - | - |
| Mouse Expression Set 430 | 1430574_at | - | - |
| Murine Genome U74v2 Set | 94971_PM_at | - | - |
| Murine Genome U74v2 Set | 94971_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1430574_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1430574_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 101770170 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2483145 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1243616 | - | - |

Cdkn3
Genomic Map