Assay Details
| Gene Symbol: Clasrp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016680.5 | NP_057889.3 | 4 - 5 | 393 | 72 | |
| NR_125832.1 | 4 - 5 | 393 | 72 | |||
| NR_125833.1 | 4 - 5 | 393 | 72 | |||
| NR_125834.1 | 4 - 5 | 393 | 72 | |||
| NR_125835.1 | 4 - 5 | 355 | 72 | |||
| GenBank mRNA | AB080582.1 | - | 3 - 4 | 256 | 72 | |
| AB080583.1 | - | 3 - 4 | 256 | 72 | ||
| AF042799.1 | - | 4 - 5 | 358 | 72 | ||
| AK009334.1 | - | 4 - 5 | 344 | 72 | ||
| AK159443.1 | - | 4 - 5 | 362 | 72 | ||
| BC010728.1 | - | 4 - 5 | 344 | 72 | ||
| BC051916.1 | - | 4 - 5 | 368 | 5707517 | 72 | |
| BC060092.1 | - | 4 - 5 | 366 | 72 | ||
| BC079543.1 | - | 4 - 5 | 356 | 72 | ||
| BC106129.1 | - | 4 - 5 | 372 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Clasrp
Genomic Map