Assay Details
| Gene Symbol: Krit1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030675.3 | NP_109600.2 | 15 - 16 | 2348 | 124 | |
| NR_033173.1 | 16 - 17 | 2595 | 124 | |||
| GenBank mRNA | AF306509.1 | - | 15 - 16 | 2043 | 124 | |
| AK085107.1 | - | 14 - 15 | 3769 | 124 | ||
| AK144667.1 | - | 15 - 16 | 2341 | 124 | ||
| AY464945.1 | - | 14 - 15 | 2030 | 124 | ||
| BC016087.1 | - | 15 - 16 | 2296 | 122 | ||
| BC044820.1 | - | 15 - 16 | 2237 | 124 | ||
| BC054819.1 | - | 16 - 17 | 2587 | 124 | ||
| BC141042.1 | - | 15 - 16 | 2192 | 9055654 | 124 | |
More Information
Set Membership: |
|

Krit1
Genomic Map