Assay Details
| Gene Symbol: Krit1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170552.1 | NP_001164023.1 | 6 - 7 | 1100 | 95 | |
| NM_030675.3 | NP_109600.2 | 6 - 7 | 1054 | 95 | ||
| NR_033173.1 | 6 - 7 | 1153 | 95 | |||
| GenBank mRNA | AF306509.1 | - | 6 - 7 | 767 | 95 | |
| AF310134.1 | - | 6 - 7 | 1030 | 95 | ||
| AK039264.1 | - | 1075 | 95 | |||
| AK085107.1 | - | 6 - 7 | 1082 | 95 | ||
| AK144667.1 | - | 6 - 7 | 1036 | 95 | ||
| AY328895.1 | - | 5 - 6 | 736 | 95 | ||
| AY464945.1 | - | 5 - 6 | 736 | 95 | ||
| BC016087.1 | - | 6 - 7 | 1004 | 95 | ||
| BC044820.1 | - | 5 - 6 | 795 | 95 | ||
| BC054819.1 | - | 6 - 7 | 1145 | 95 | ||
| BC085252.1 | - | 6 - 7 | 860 | 95 | ||
| BC141042.1 | - | 6 - 7 | 898 | 9055654 | 95 | |
More Information
Updated Mapping Information
Set Membership: |
|

Krit1
Genomic Map