Assay Details
| Gene Symbol: Krit1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170552.1 | NP_001164023.1 | 9 - 10 | 1512 | 114 | |
| NM_030675.3 | NP_109600.2 | 9 - 10 | 1466 | 114 | ||
| NR_033173.1 | 10 - 11 | 1713 | 114 | |||
| GenBank mRNA | AF306509.1 | - | 9 - 10 | 1161 | 114 | |
| AF310134.1 | - | 9 - 10 | 1442 | 114 | ||
| AK039264.1 | - | 1343 | 114 | |||
| AK085107.1 | - | 9 - 10 | 1494 | 114 | ||
| AK144667.1 | - | 9 - 10 | 1448 | 114 | ||
| AY328895.1 | - | 8 - 9 | 1148 | 114 | ||
| AY464945.1 | - | 8 - 9 | 1148 | 114 | ||
| BC016087.1 | - | 9 - 10 | 1416 | 114 | ||
| BC044820.1 | - | 9 - 10 | 1355 | 114 | ||
| BC054819.1 | - | 10 - 11 | 1705 | 114 | ||
| BC085252.1 | - | 9 - 10 | 1272 | 114 | ||
| BC141042.1 | - | 9 - 10 | 1310 | 9055654 | 114 | |
More Information
Updated Mapping Information
Set Membership: |
|

Krit1
Genomic Map