Assay Details
| Gene Symbol: Ube2e2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144839.1 | NP_659088.1 | 1 - 2 | 44 | 58 | |
| GenBank mRNA | BC016265.1 | - | 1 - 2 | 44 | 4923869 | 58 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC016265.1, NM_144839.1 transcripts.
Set Membership: |
|

Ube2e2
Genomic Map