Assay Details
| Gene Symbol: Rab10os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_015551.1 | 4 - 5 | 422 | 114 | ||
| GenBank mRNA | AK005738.1 | - | 4 - 5 | 375 | 114 | |
| AK007702.1 | - | 4 - 5 | 423 | 114 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Downstream |
RAB10, member RAS oncogene family |
Rab10 | AW107754 | 5.9kb | Anti-sense | Chr. 12 - 3247428 - 3309969 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Rab10os
Genomic Map