Assay Details
| Gene Symbol: Fam228a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK015158.1 | - | 2 - 3 | 158 | 154 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK015158.1 transcripts.
Set Membership: |
|

Fam228a
Genomic Map