Assay Details
| Gene Symbol: Ptpn14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008976.2 | NP_033002.2 | 16 - 17 | 3581 | 77 | |
| GenBank mRNA | AF170902.1 | - | 15 - 16 | 2232 | 77 | |
| AF170903.1 | - | 15 - 16 | 2128 | 77 | ||
| AK156480.1 | - | 16 - 17 | 3327 | 77 | ||
| CK780322.1 | - | 1 - 2 | 143 | 77 | ||
| D31842.1 | - | 15 - 16 | 3178 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ptpn14
Genomic Map