Assay Details
| Gene Symbol: Enah | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083120.2 | NP_001076589.1 | 4 - 5 | 880 | 61 | |
| NM_001083121.2 | NP_001076590.1 | 4 - 5 | 880 | 61 | ||
| NM_008680.3 | NP_032706.2 | 5 - 6 | 892 | 61 | ||
| NM_010135.3 | NP_034265.2 | 5 - 6 | 937 | 61 | ||
| GenBank mRNA | BC062927.1 | - | 4 - 5 | 499 | 6851217 | 61 |
| U72520.1 | - | 4 - 5 | 573 | 61 | ||
| U72521.1 | - | 4 - 5 | 573 | 61 | ||
| U72522.1 | - | 5 - 6 | 585 | 61 | ||
| U72523.1 | - | 5 - 6 | 630 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Enah
Genomic Map