Assay Details
| Gene Symbol: Prkacb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164200.1 | NP_001157672.1 | 1 - 2 | 264 | 92 | |
| GenBank mRNA | AK150122.1 | - | 1 - 2 | 265 | 92 | |
| AK150134.1 | - | 1 - 2 | 265 | 92 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prkacb
Genomic Map