Assay Details
| Gene Symbol: Gbp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289492.1 | NP_001276421.1 | 9 - 10 | 1740 | 83 | |
| NM_001289493.1 | NP_001276422.1 | 8 - 9 | 1565 | 83 | ||
| NM_018734.3 | NP_061204.3 | 8 - 9 | 1602 | 83 | ||
| GenBank mRNA | AK132266.1 | - | 8 - 9 | 1601 | 83 | |
| BC019195.1 | - | 9 - 10 | 1651 | 5036920 | 83 | |
| U44731.1 | - | 8 - 9 | 1454 | 83 | ||
More Information
Set Membership: |
|

Gbp3
Genomic Map