Assay Details
| Gene Symbol: Sh3glb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282037.1 | NP_001268966.1 | 6 - 7 | 895 | 99 | |
| GenBank mRNA | AF272946.1 | - | 6 - 7 | 883 | 99 | |
More Information
Set Membership: |
|

Sh3glb1
Genomic Map