Assay Details
| Gene Symbol: Sh3glb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282037.1 | NP_001268966.1 | 7 - 8 | 916 | 98 | |
| GenBank mRNA | AF272946.1 | - | 7 - 8 | 904 | 98 | |
More Information
Set Membership: |
|

Sh3glb1
Genomic Map