Assay Details
| Gene Symbol: Ptbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019550.2 | NP_062423.1 | 12 - 13 | 1572 | 69 | |
| GenBank mRNA | AF095718.2 | - | 12 - 13 | 1442 | 69 | |
| AK013734.1 | - | 1 - 2 | 135 | 69 | ||
| AK033964.1 | - | 5 - 6 | 1979 | 69 | ||
| AK135738.1 | - | 12 - 13 | 1575 | 69 | ||
| AK137348.1 | - | 12 - 13 | 1451 | 69 | ||
| BC010255.1 | - | 12 - 13 | 1458 | 3709255 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ptbp2
Genomic Map