Assay Details
| Gene Symbol: Ptbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019550.2 | NP_062423.1 | 4 - 5 | 473 | 118 | |
| GenBank mRNA | AF095718.2 | - | 4 - 5 | 343 | 118 | |
| AK041655.1 | - | 4 - 5 | 372 | 118 | ||
| AK050624.1 | - | 4 - 5 | 354 | 118 | ||
| AK050650.1 | - | 4 - 5 | 354 | 118 | ||
| AK135738.1 | - | 4 - 5 | 474 | 118 | ||
| AK137348.1 | - | 4 - 5 | 352 | 118 | ||
| BC010255.1 | - | 4 - 5 | 359 | 3709255 | 118 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ptbp2
Genomic Map