Assay Details
| Gene Symbol: Cfi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007686.2 | NP_031712.2 | 5 - 6 | 864 | 77 | |
| GenBank mRNA | BC150751.1 | - | 5 - 6 | 857 | 9087662 | 77 |
| CN691735.1 | - | 2 - 3 | 316 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cfi
Genomic Map