Assay Details
| Gene Symbol: Cfi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007686.2 | NP_031712.2 | 7 - 8 | 1001 | 104 | |
| GenBank mRNA | BC150751.1 | - | 7 - 8 | 994 | 9087662 | 104 |
| CN691735.1 | - | 4 - 5 | 453 | 104 | ||
| U47810.1 | - | 7 - 8 | 1004 | 104 | ||
More Information
Set Membership: |
|

Cfi
Genomic Map