Assay Details
| Gene Symbol: Cfi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007686.2 | NP_031712.2 | 8 - 9 | 1020 | 104 | |
| GenBank mRNA | BC150751.1 | - | 8 - 9 | 1013 | 9087662 | 104 |
| CN691735.1 | - | 5 - 6 | 472 | 104 | ||
| U47810.1 | - | 8 - 9 | 1023 | 104 | ||
More Information
Set Membership: |
|

Cfi
Genomic Map