Assay Details
| Gene Symbol: Ppp3cc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK015002.1 | - | 4 - 5 | 705 | 78 | |
| AK133393.1 | - | 4 - 5 | 726 | 78 | ||
| BC046617.1 | - | 4 - 5 | 531 | 78 | ||
| BC141079.1 | - | 4 - 5 | 546 | 9055691 | 78 | |
| BC145347.1 | - | 3 - 4 | 421 | 78 | ||
| M81475.1 | - | 4 - 5 | 699 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ppp3cc
Genomic Map