Assay Details
| Gene Symbol: Rgs7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199003.1 | NP_001185932.1 | 12 - 13 | 1167 | 140 | |
| NM_011880.3 | NP_036010.2 | 12 - 13 | 1167 | 140 | ||
| GenBank mRNA | AF011360.1 | - | 12 - 13 | 1012 | 140 | |
| BC051133.1 | - | 12 - 13 | 1122 | 6588901 | 140 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rgs7
Genomic Map