Assay Details
| Gene Symbol: Esd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285423.1 | NP_001272352.1 | 7 - 8 | 833 | 77 | |
| NM_016903.5 | NP_058599.1 | 7 - 8 | 700 | 77 | ||
| GenBank mRNA | AB025408.1 | - | 7 - 8 | 675 | 77 | |
| AK007527.1 | - | 7 - 8 | 688 | 77 | ||
| AK010683.1 | - | 7 - 8 | 705 | 77 | ||
| AK021209.1 | - | 7 - 8 | 628 | 77 | ||
| AK079131.1 | - | 7 - 8 | 653 | 77 | ||
| AK088115.1 | - | 7 - 8 | 748 | 77 | ||
| AK163074.1 | - | 7 - 8 | 629 | 77 | ||
| BC046766.1 | - | 7 - 8 | 625 | 5694094 | 77 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Esd
Genomic Map