Assay Details
| Gene Symbol: Cab39l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026908.3 | NP_081184.3 | 5 - 6 | 554 | 99 | |
| GenBank mRNA | AK005323.1 | - | 553 | 99 | ||
| AK013161.1 | - | 4 - 5 | 413 | 99 | ||
| AK013205.1 | - | 4 - 5 | 549 | 99 | ||
| AK030474.1 | - | 5 - 6 | 568 | 99 | ||
| AK053642.1 | - | 5 - 6 | 555 | 99 | ||
| AK076758.1 | - | 5 - 6 | 555 | 99 | ||
| AK076867.1 | - | 5 - 6 | 575 | 99 | ||
| BC016128.1 | - | 3 - 4 | 521 | 4911640 | 99 | |
| BC016546.1 | - | 5 - 6 | 538 | 3595339 | 99 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cab39l
Genomic Map