Assay Details
| Gene Symbol: Diap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019670.1 | NP_062644.1 | 13 - 14 | 1418 | 90 | |
| GenBank mRNA | AF094519.1 | - | 13 - 14 | 1418 | 90 | |
| AK137073.1 | - | 12 - 13 | 1507 | 90 | ||
| AK138694.1 | - | 13 - 14 | 1448 | 90 | ||
| AK161737.1 | - | 12 - 13 | 1436 | 90 | ||
| AK166882.1 | - | 13 - 14 | 1474 | 90 | ||
| AK220493.1 | - | 13 - 14 | 1482 | 90 | ||
| BC003250.1 | - | 12 - 13 | 1452 | 90 | ||
| BC028920.1 | - | 6 - 7 | 719 | 4008236 | 90 | |
| BC085191.1 | - | 13 - 14 | 1528 | 30693873 | 90 | |
| BC086779.1 | - | 13 - 14 | 1493 | 90 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Diap3
Genomic Map