Assay Details
| Gene Symbol: Diap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019670.1 | NP_062644.1 | 16 - 17 | 1900 | 133 | |
| GenBank mRNA | AF094519.1 | - | 16 - 17 | 1900 | 133 | |
| AK137073.1 | - | 15 - 16 | 1989 | 133 | ||
| AK138694.1 | - | 16 - 17 | 1930 | 133 | ||
| AK161737.1 | - | 15 - 16 | 1918 | 133 | ||
| AK166882.1 | - | 16 - 17 | 1956 | 133 | ||
| AK220493.1 | - | 16 - 17 | 1964 | 133 | ||
| BC003250.1 | - | 15 - 16 | 1934 | 133 | ||
| BC028920.1 | - | 9 - 10 | 1201 | 4008236 | 133 | |
| BC085191.1 | - | 16 - 17 | 2010 | 30693873 | 133 | |
| BC086779.1 | - | 16 - 17 | 1975 | 133 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Diap3
Genomic Map