Assay Details
| Gene Symbol: Diap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019670.1 | NP_062644.1 | 17 - 18 | 2001 | 154 | |
| GenBank mRNA | AF094519.1 | - | 17 - 18 | 2001 | 154 | |
| AK138694.1 | - | 17 - 18 | 2031 | 154 | ||
| AK161737.1 | - | 16 - 17 | 2019 | 154 | ||
| AK166882.1 | - | 17 - 18 | 2057 | 154 | ||
| AK220493.1 | - | 17 - 18 | 2065 | 154 | ||
| BC085191.1 | - | 17 - 18 | 2111 | 30693873 | 154 | |
| BC086779.1 | - | 17 - 18 | 2076 | 154 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Diap3
Genomic Map